CAS RN 470-67-7
IUPAC Name:
1-methyl-4-(propan-2-yl)-7-oxabicyclo[2.2.1]heptane
Synonyms:
1,4-cineol isocineole
Molecular Weight (g/mol):
154.25
Molecular Formula:
C10H18O
InChi Key:
RFFOTVCVTJUTAD-UHFFFAOYSA-N
SMILES:
CC(C)C12CCC(C)(CC1)O2
1
–
1
of
1
results
1
1,4-Cineole, 85%
CAS: 470-67-7 Molecular Formula: C10H18O Molecular Weight (g/mol): 154.25 InChI Key: RFFOTVCVTJUTAD-UHFFFAOYSA-N Synonym: 1,4-cineole,isocineole,1,4-cineol,1,4-epoxy-p-menthane,isocineple,1,4-cineole natural,p-menthane, 1,4-epoxy,unii-b55jtu839b,fema no. 3658 PubChem CID: 10106 ChEBI: CHEBI:80788 IUPAC Name: 1-methyl-4-propan-2-yl-7-oxabicyclo[2.2.1]heptane SMILES: CC(C)C12CCC(O1)(CC2)C