missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cineole, 85%

Product Code. 15412162 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15412162 100 g 100g
15402162 25 g 25g
15422162 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15412162 Supplier Thermo Scientific Acros Supplier No. 456061000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 470-67-7 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 470-67-7
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
InChI Key RFFOTVCVTJUTAD-UHFFFAOYSA-N
Synonym 1,4-cineole, isocineole, 1,4-cineol, 1,4-epoxy-p-menthane, isocineple, 1,4-cineole natural, p-menthane, 1,4-epoxy, unii-b55jtu839b, fema no. 3658
PubChem CID 10106
ChEBI CHEBI:80788
IUPAC Name 1-methyl-4-propan-2-yl-7-oxabicyclo[2.2.1]heptane
SMILES CC(C)C12CCC(O1)(CC2)C

Specifications

Melting Point -46.0°C
Density 0.8870g/mL
Boiling Point 65°C (21.0 hPa)
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 80% min. (GC)
Quantity 100 g
Specific Gravity 0.887
Formula Weight 154.25
Percent Purity 85%
Chemical Name or Material 1,4-Cineole
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Cineole
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.