CAS RN 4704-99-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
alpha,alpha,alpha-Tris(hydroxymethyl)toluene, TRC
CAS: 4704-99-8 Molecular Formula: C10 H14 O3 Molecular Weight (g/mol): 182.22 Synonym: 2-Hydroxymethyl-2-phenylpropane-1,3-diol,β,β-Bis(hydroxymethyl)-benzeneethanol IUPAC Name: 2-(hydroxymethyl)-2-phenylpropane-1,3-diol SMILES: OCC(CO)(CO)c1ccccc1