missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha-Tris(hydroxymethyl)toluene, TRC

Product Code. 30457294
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457294 10 g 10g
30456491 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30457294 Supplier Toronto Research Chemicals Supplier No. TRCT88491010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4704-99-8
Molecular Formula C10 H14 O3
Molecular Weight (g/mol) 182.22
Synonym 2-Hydroxymethyl-2-phenylpropane-1,3-diol, β,β-Bis(hydroxymethyl)-benzeneethanol
IUPAC Name 2-(hydroxymethyl)-2-phenylpropane-1,3-diol
SMILES OCC(CO)(CO)c1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H14O3/c11-6-10(7-12,8-13)9-4-2-1-3-5-9/h1-5,11-13H,6-8H2
Formula Weight 182.0943
Chemical Name or Material alpha,alpha,alpha-Tris(hydroxymethyl)toluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.