CAS RN 4727-31-5
IUPAC Name:
2-({[1,1'-biphenyl]-4-yl}carbamoyl)benzoic acid
Synonyms:
2-({[1,1'-biphenyl]-4-yl}carbamoyl)benzoic acid
Molecular Weight (g/mol):
317.34
Molecular Formula:
C20H15NO3
InChi Key:
SLUINPGXGFUMLL-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=CC=C1C(=O)NC1=CC=C(C=C1)C1=CC=CC=C1
1
–
3
of
3
results
1
Thermo Scientific Chemicals Kartogenin
CAS: 4727-31-5 Molecular Formula: C20H15NO3 Molecular Weight (g/mol): 317.34 InChI Key: SLUINPGXGFUMLL-UHFFFAOYSA-N IUPAC Name: 2-({[1,1'-biphenyl]-4-yl}carbamoyl)benzoic acid SMILES: OC(=O)C1=CC=CC=C1C(=O)NC1=CC=C(C=C1)C1=CC=CC=C1
2
3
Kartogenin, TRC
CAS: 4727-31-5 Molecular Formula: C20 H15 N O3 Molecular Weight (g/mol): 317.34 Synonym: 2-[([1,1'-Biphenyl]-4-ylamino)carbonyl]benzoic Acid,2-[(Biphenyl-4-yl)carbamoyl]benzoic Acid,4'-Phenylphthalanilic Acid,KGN; IUPAC Name: 2-[(4-phenylphenyl)carbamoyl]benzoic acid SMILES: OC(=O)c1ccccc1C(=O)Nc2ccc(cc2)c3ccccc3