missing translation for 'onlineSavingsMsg'
Learn More

Kartogenin, TRC

Product Code. 30137020
Change view
Click to view available options
Quantity:
100 mg
10 g
Unit Size:
100mg
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137020 10 g 10g
30003400 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137020 Supplier Toronto Research Chemicals Supplier No. TRCK13980010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4727-31-5
Molecular Formula C20 H15 N O3
Molecular Weight (g/mol) 317.34
Synonym 2-[([1,1'-Biphenyl]-4-ylamino)carbonyl]benzoic Acid, 2-[(Biphenyl-4-yl)carbamoyl]benzoic Acid, 4'-Phenylphthalanilic Acid, KGN;
IUPAC Name 2-[(4-phenylphenyl)carbamoyl]benzoic acid
SMILES OC(=O)c1ccccc1C(=O)Nc2ccc(cc2)c3ccccc3

Specifications

Quantity 10 g
InChI Formula InChI=1S/C20H15NO3/c22-19(17-8-4-5-9-18(17)20(23)24)21-16-12-10-15(11-13-16)14-6-2-1-3-7-14/h1-13H,(H,21,22)(H,23,24)
Formula Weight 317.1052
Chemical Name or Material Kartogenin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.