accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 473917-15-6

OFOFFF

CAS RN 473917-15-6

IUPAC Name: 3-fluoro-4-(trifluoromethoxy)benzaldehyde
Synonyms: 3-fluoro-4-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 208.11
Molecular Formula: C8H4F4O2
InChi Key: RQLRUBHAAVGRPW-UHFFFAOYSA-N
SMILES: FC1=C(OC(F)(F)F)C=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results