accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOHOOHOOH

CAS RN 475-25-2

IUPAC Name: 5,6,10,15-tetrahydroxy-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-1(17),2,4,6,12,15-hexaen-14-one
Synonyms: hematein
Molecular Weight (g/mol): 300.27
Molecular Formula: C16H12O6
InChi Key: HLUCICHZHWJHLL-UHFFFAOYNA-N
SMILES: OC1=CC=C2C3=C4C=C(O)C(=O)C=C4CC3(O)COC2=C1O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)