accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOCH3OH3C(R)(S)(R)OO(R)OHOO

CAS RN 477-47-4

IUPAC Name: (10R,11S,15R,16R)-16-hydroxy-10-(3,4,5-trimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one
Synonyms: (10R,11S,15R,16R)-16-hydroxy-10-(3,4,5-trimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one
Molecular Weight (g/mol): 414.41
Molecular Formula: C22H22O8
InChi Key: YJGVMLPVUAXIQN-HAEOHBJNSA-N
SMILES: COC1=CC(=CC(OC)=C1OC)[C@H]1[C@H]2[C@H](COC2=O)[C@@H](O)C2=CC3=C(OCO3)C=C12
Quantity 
  • (1)
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results