accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHN

CAS RN 4771-50-0

IUPAC Name: 7-methyl-1H-indole-3-carbaldehyde
Synonyms: 7-methyl-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: KTUFZHVVJBHGKZ-UHFFFAOYSA-N
SMILES: CC1=C2NC=C(C=O)C2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results