accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4792-58-9

IUPAC Name: ethyl 5-methoxy-1H-indole-2-carboxylate
Synonyms: ethyl 5-methoxy-1H-indole-2-carboxylate
Molecular Weight (g/mol): 219.24
Molecular Formula: C12H13NO3
InChi Key: NPIUAXNFAUGNHP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC(OC)=CC=C2N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results