accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 479411-92-2

IUPAC Name: 2-[3-bromo-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[3-bromo-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 350.97
Molecular Formula: C13H15BBrF3O2
InChi Key: CNCVCSYYPDHXMW-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CC(Br)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results