missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(trifluoromethyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 15489285
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15489285 250 mg 250mg
15479285 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15489285 Supplier Thermo Scientific Alfa Aesar Supplier No. H51713.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as a active pharmaceutical ingredient.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479411-92-2
Molecular Formula C13H15BBrF3O2
Molecular Weight (g/mol) 350.97
MDL Number MFCD12405333
InChI Key CNCVCSYYPDHXMW-UHFFFAOYSA-N
Synonym 2-3-bromo-5-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-bromo-5-trifluoromethyl benzeneboronic acid pinacol ester, 2-3-bromo-5-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,
PubChem CID 11314119
IUPAC Name 2-[3-bromo-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)Br)C(F)(F)F

Specifications

Melting Point 50°C to 52°C
Quantity 250 mg
Formula Weight 350.97
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(trifluoromethyl)benzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.