accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHHOHOO

CAS RN 480-64-8

IUPAC Name: 2,4-dihydroxy-6-methylbenzoic acid
Synonyms: orsellinic acid o-orsellinic acid orsellic acid orsellinsaeure
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: AMKYESDOVDKZKV-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC(O)=C1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results