missing translation for 'onlineSavingsMsg'
Learn More

Orsellinic Acid, TRC

Product Code. 30421193
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421193 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30421193 Supplier Toronto Research Chemicals Supplier No. TRCO69760850MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Orsellinic Acid
CAS 480-64-8
Molecular Formula C8H8O4
Quantity 50 mg
Synonym Orsellinic Acid, 2,4-Dihydroxy-6-methylbenzoic acid, 2,4-Dihydroxy-6-methylbenzoic Acid, 2,4-Dihydroxy-6-methylbenzenecarboxylic Acid, 4,6-Dihydroxy-2-methylbenzoic Acid, 6-Methyl-β-resorcylic Acid, Orcinolcarboxylic Acid, o-Orsellinic Acid
InChI Formula InChI=1S/C8H8O4/c1-4-2-5(9)3-6(10)7(4)8(11)12/h2-3,9-10H,1H3,(H,11,12)
SMILES Cc1cc(O)cc(O)c1C(=O)O
IUPAC Name 2,4-dihydroxy-6-methylbenzoic acid
Molecular Weight (g/mol) 168.15
Formula Weight 168.0423
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.