accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 480-96-6

IUPAC Name: 2,1,3-benzoxadiazol-1-ium-1-olate
Synonyms: 2,1,3-benzoxadiazol-1-ium-1-olate
Molecular Weight (g/mol): 136.11
Molecular Formula: C6H4N2O2
InChi Key: OKEAMBAZBICIFP-UHFFFAOYSA-N
SMILES: [O-][N+]1=C2C=CC=CC2=NO1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)