missing translation for 'onlineSavingsMsg'
Learn More

Benzofuroxan, 98%, BLD Pharm™

Product Code. 31006042
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31006042 500 g 500g
31001649 1 kg 1kg
30993819 100 g 100g
30990980 5 g 5g
30999616 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31006042 Supplier BLD Pharm Supplier No. BD115767500g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 480-96-6
Molecular Formula C6H4N2O2
MDL Number MFCD00005807
Quantity 500 g
SMILES [O-][N+]1=C2C=CC=CC2=NO1
Molecular Weight (g/mol) 136.11
Percent Purity 98%
Chemical Name or Material Benzofuroxan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.