accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4815-37-6

IUPAC Name: ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
Synonyms: ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
Molecular Weight (g/mol): 261.34
Molecular Formula: C14H15NO2S
InChi Key: SXONDAWSQJWZEO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC(C)=C1C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)