missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate, 97%

Product Code. 15489335
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15489335 5 g 5g
15479335 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15489335 Supplier Alfa Aesar™ Supplier No. H51799.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4815-37-6
Molecular Formula C14H15NO2S
Molecular Weight (g/mol) 261.339
MDL Number MFCD01763475
InChI Key SXONDAWSQJWZEO-UHFFFAOYSA-N
Synonym 2-amino-5-methyl-4-phenyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylicacid, 2-amino-5-methyl-4-phenyl-, ethyl ester, maybridge1_000503, upcmld-dp080, upcmld-dp080:001, ethyl 2-amino-5-methyl-4-phenyl-3-thiophenecarboxylate, 3-thiophenecarboxylic acid,2-amino-5-methyl-4-phen, 2-amino-4-phenyl-5-methylthiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-5-methyl-4-phenyl-, ethyl ester
PubChem CID 693902
IUPAC Name ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC(=C1C2=CC=CC=C2)C)N

Specifications

Quantity 5 g
Formula Weight 261.34
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.