accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3HOOOHO

CAS RN 4839-46-7

IUPAC Name: 3,3-dimethylpentanedioic acid
Synonyms: β,β-dimethylglutaric acid b,b-dimethylglutarate b,b-dimethylglutaric acid
Molecular Weight (g/mol): 160.17
Molecular Formula: C7H12O4
InChi Key: DUHQIGLHYXLKAE-UHFFFAOYSA-N
SMILES: CC(C)(CC(O)=O)CC(O)=O
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)