missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylglutaric Acid, TRC

Product Code. 30385205
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385205 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385205 Supplier Toronto Research Chemicals Supplier No. TRCD47391510G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,3-Dimethylglutaric Acid
CAS 4839-46-7
Molecular Formula C7H12O4
Quantity 10 g
Synonym 3,3-Dimethyl-pentanedioic Acid, beta,beta-Dimethyl-glutaric Acid, 3,3-Dimethylpentanedioic Acid, NSC 14987, NSC 49114, beta,beta-Dimethylglutaric Acid
InChI Formula InChI=1S/C7H12O4/c1-7(2,3-5(8)9)4-6(10)11/h3-4H2,1-2H3,(H,8,9)(H,10,11)
SMILES CC(C)(CC(=O)O)CC(=O)O
IUPAC Name 3,3-dimethylpentanedioic acid
Molecular Weight (g/mol) 160.17
Formula Weight 160.0736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.