accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3OH3C

CAS RN 487-68-3

IUPAC Name: 2,4,6-trimethylbenzaldehyde
Synonyms: benzaldehyde, 2,4,6-trimethyl-
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: HIKRJHFHGKZKRI-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C=O)C(C)=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)