missing translation for 'onlineSavingsMsg'
Learn More

Mesitaldehyde, TRC

Product Code. 17943595
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17943595 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17943595 Supplier Toronto Research Chemicals Supplier No. TRCM2584005G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 487-68-3
Molecular Formula C10 H12 O
Quantity 5 g
Synonym 2,4,6-Trimethylbenzaldehyde (ACI), 2-Mesitylenecarboxaldehyde, Mesitaldehyde, Mesitylaldehyde, Mesitylenecarboxaldehyde, NSC 29094, 2,4,6-Trimethylbenzaldehyde
InChI Formula InChI=1S/C10H12O/c1-7-4-8(2)10(6-11)9(3)5-7/h4-6H,1-3H3
SMILES Cc1cc(C)c(C=O)c(C)c1
IUPAC Name 2,4,6-trimethylbenzaldehyde
Molecular Weight (g/mol) 148.2
Formula Weight 148.0888
Chemical Name or Material Mesitaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.