accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 488-86-8

IUPAC Name: 5-hydroxycyclopentane-1,2,3,4-tetrone
Synonyms: croconic acid
Molecular Weight (g/mol): 142.07
Molecular Formula: C5H2O5
InChi Key: WYNWSVZYZJTUNK-UHFFFAOYSA-N
SMILES: OC1C(=O)C(=O)C(=O)C1=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (5)
  • (1)
  • (6)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)