CAS RN 492-88-6
IUPAC Name:
3-ethoxy-2-hydroxybenzaldehyde
Synonyms:
3-ethoxy-2-hydroxybenzaldehyde
Molecular Weight (g/mol):
166.18
Molecular Formula:
C9H10O3
InChi Key:
OFQBYHLLIJGMNP-UHFFFAOYSA-N
SMILES:
CCOC1=CC=CC(C=O)=C1O
1
–
1
of
1
results
1
3-Ethoxysalicylaldehyde, 97%
CAS: 492-88-6 Molecular Formula: C9H10O3 Molecular Weight (g/mol): 166.176 MDL Number: MFCD00003323 InChI Key: OFQBYHLLIJGMNP-UHFFFAOYSA-N Synonym: 3-ethoxysalicylaldehyde,benzaldehyde, 3-ethoxy-2-hydroxy,novovanillin,o-ethylvanillin,salicylaldehyde, 3-ethoxy,unii-hb4h48mkni,2-hydroxy-3-ethoxybenzaldehyde,hb4h48mkni,3-ethoxy-2-hydroxy-benzaldehyde,pubchem3123 PubChem CID: 68117 IUPAC Name: 3-ethoxy-2-hydroxybenzaldehyde SMILES: CCOC1=CC=CC(=C1O)C=O