accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 495-71-6

OO

CAS RN 495-71-6

IUPAC Name: 1,4-diphenylbutane-1,4-dione
Synonyms: 1,4-diphenylbutane-1,4-dione
Molecular Weight (g/mol): 238.29
Molecular Formula: C16H14O2
InChi Key: OSWWFLDIIGGSJV-UHFFFAOYSA-N
SMILES: O=C(CCC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results