accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COO(S)(S)(R)OH(S)(S)H3C

CAS RN 4956-37-0

IUPAC Name: (1S,3aS,3bR,9bS,11aS)-7-hydroxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl heptanoate
Synonyms: estradiol enanthate
Molecular Weight (g/mol): 384.56
Molecular Formula: C25H36O3
InChi Key: RFWTZQAOOLFXAY-BZDYCCQFSA-N
SMILES: CCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(O)=CC=C4[C@H]3CC[C@]12C
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results