Learn More
Estradiol Enanthate, Mikromol™
Description
Estradiol Enanthate
Specifications
Specifications
| Chemical Name or Material | Estradiol Enanthate |
| CAS | 4956-37-0 |
| Analyte or Component Names | Estradiol Enanthate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H36 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Estradiol Enanthate |
| InChI Formula | InChI=1S/C25H36O3/c1-3-4-5-6-7-24(27)28-23-13-12-22-21-10-8-17-16-18(26)9-11-19(17)20(21)14-15-25(22,23)2/h9,11,16,20-23,26H,3-8,10,12-15H2,1-2H3/t20-,21-,22+,23+,25+/m1/s1 |
| SMILES | CCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C |
| IUPAC Name | [(8R,9S,13S,14S,17S)-3-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] heptanoate |
| Show More |