accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHOCH3

CAS RN 496-77-5

IUPAC Name: 5-hydroxyoctan-4-one
Synonyms: 5-hydroxyoctan-4-one
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: BVEYJWQCMOVMAR-UHFFFAOYNA-N
SMILES: CCCC(O)C(=O)CCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results