accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 497-38-1

O

CAS RN 497-38-1

IUPAC Name: bicyclo[2.2.1]heptan-2-one
Synonyms: norcamphor
Molecular Weight (g/mol): 110.16
Molecular Formula: C7H10O
InChi Key: KPMKEVXVVHNIEY-UHFFFAOYNA-N
SMILES: O=C1CC2CCC1C2
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)