Learn More
Norcamphor, 98+%
Description
Norcamphor is used as a building block in organic synthesis. It is also used as a precursor to norborneols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Norcamphor is used as a building block in organic synthesis. It is also used as a precursor to norborneols.
Solubility
Insoluble in water. Soluble in methanol.
Notes
Incompatible with strong oxidizing agents. Heat and light sensitive.
Specifications
Specifications
| Melting Point | 92°C to 97°C |
| Boiling Point | 172°C to 174°C |
| Flash Point | 33°C (91°F) |
| Quantity | 25 g |
| UN Number | UN1325 |
| Beilstein | 1209657 |
| Solubility Information | Insoluble in water. Soluble in methanol. |
| Formula Weight | 110.16 |
| Percent Purity | ≥98% |
| Chemical Name or Material | Norcamphor |
Safety and Handling
- Norcamphor
- Warning
- Flammable solid Category 2
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- MIXTURE LIST-Contains : Bicyclo[2.2.1]heptan-2-one
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.