accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 50-14-6

IUPAC Name: (1S,3E)-3-{2-[(1R,3aS,4E,7aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol
Synonyms: β-ol sorex C.R. ergocalciferolo vio D calciferolum osteil ergocalciferol oil HI-deratol uvesterol D ergosterol irradiated
Molecular Weight (g/mol): 396.66
Molecular Formula: C28H44O
InChi Key: MECHNRXZTMCUDQ-VLOQVYPSSA-N
SMILES: CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C
Molecular Weight (g/mol) 
  • (2)
  • (9)
Quantity 
  • (3)
  • (1)
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (9)
  • (9)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)