accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCH3

CAS RN 502-56-7

IUPAC Name: nonan-5-one
Synonyms: 5-nonanone
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: WSGCRAOTEDLMFQ-UHFFFAOYSA-N
SMILES: CCCCC(=O)CCCC
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)