missing translation for 'onlineSavingsMsg'
Learn More

5-Nonanone, 98%

Product Code. 11489050
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489050 500 g 500g
11479050 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489050 Supplier Thermo Scientific Alfa Aesar Supplier No. A13722.36
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 502-56-7 | C9H18O | 142.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-56-7
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00009484
InChI Key WSGCRAOTEDLMFQ-UHFFFAOYSA-N
Synonym 5-nonanone, dibutyl ketone, butyl ketone, 5-oxononane, di-n-butyl ketone, unii-v8b2y1bz3s, hsdb 921, v8b2y1bz3s, n-c4h9 2co, dibutylketon
PubChem CID 10405
IUPAC Name nonan-5-one
SMILES CCCCC(=O)CCCC

Specifications

Melting Point -50°C
Density 0.824
Boiling Point 186°C to 187°C
Flash Point 65°C (149°F)
Refractive Index 1.419
Quantity 500 g
Beilstein 1743583
Formula Weight 142.24
Percent Purity 98%
Chemical Name or Material 5-Nonanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.