CAS RN 50262-48-1
IUPAC Name:
4-formylphenyl propanoate
Synonyms:
4-formylphenyl propanoate
Molecular Weight (g/mol):
178.19
Molecular Formula:
C10H10O3
InChi Key:
NKXPJXVTMWLHBC-UHFFFAOYSA-N
SMILES:
CCC(=O)OC1=CC=C(C=O)C=C1
1
–
1
of
1
results
1
4-Propionyloxybenzaldehyde, 95%, Thermo Scientific™
CAS: 50262-48-1 Molecular Formula: C10H10O3 Molecular Weight (g/mol): 178.187 MDL Number: MFCD00043522 InChI Key: NKXPJXVTMWLHBC-UHFFFAOYSA-N Synonym: 4-formylphenyl propionate,4-propionyloxybenzaldehyde,p-propionoxybenzaldehyde,benzaldehyde, 4-1-oxopropoxy,4-n-propionyloxybenzaldehyde,p-formylphenyl propionate,acmc-20amb3,4-formylphenyl propanoate,4-formylphenyl propionate #,4-propanyloxy-benzaldehyde PubChem CID: 123519 IUPAC Name: (4-formylphenyl) propanoate SMILES: CCC(=O)OC1=CC=C(C=C1)C=O