missing translation for 'onlineSavingsMsg'
Learn More

4-Propionyloxybenzaldehyde, 95%, Thermo Scientific™

Product Code. 11342686
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342686 25 g 25g
11332686 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342686 Supplier Thermo Scientific Alfa Aesar Supplier No. L08732.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50262-48-1
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00043522
InChI Key NKXPJXVTMWLHBC-UHFFFAOYSA-N
Synonym 4-formylphenyl propionate, 4-propionyloxybenzaldehyde, p-propionoxybenzaldehyde, benzaldehyde, 4-1-oxopropoxy, 4-n-propionyloxybenzaldehyde, p-formylphenyl propionate, acmc-20amb3, 4-formylphenyl propanoate, 4-formylphenyl propionate #, 4-propanyloxy-benzaldehyde
PubChem CID 123519
IUPAC Name (4-formylphenyl) propanoate
SMILES CCC(=O)OC1=CC=C(C=C1)C=O

Specifications

Boiling Point 132°C to 136°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.53
Quantity 25 g
Beilstein 2966420
Sensitivity Air Sensitive
Formula Weight 178.19
Percent Purity 95%
Chemical Name or Material 4-Propionyloxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.