accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 503-74-2

IUPAC Name: 3-methylbutanoic acid
Synonyms: isovaleric acid fema number: 3102 isovalerianic acid isovaleric acid (natural) isovalerianic 3-methylbuttersaeure kyselina isovalerova delphinic acid isovaleriansaeure
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: GWYFCOCPABKNJV-UHFFFAOYSA-N
SMILES: CC(C)CC(O)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results