missing translation for 'onlineSavingsMsg'
Learn More

Isovaleric Acid, TRC

Product Code. 30390448
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390448 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390448 Supplier Toronto Research Chemicals Supplier No. TRCI917570100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 503-74-2
Molecular Formula C5 H10 O2
Quantity 100 g
Synonym Butanoic acid, 3-methyl- (9CI, ACI), 3-Methylbutanoic acid (ACI), Isovaleric acid (8CI), 3-Methyl-1-butanoic acid, 3-Methyl-n-butyric acid, 3-Methylbutyrate, 3-Methylbutyric acid, Acetic acid, isopropyl-, Delphinic acid, Isopentanoic acid, Isopropylacetic acid, Isovalerianic acid, NSC 62783, β-Methylbutyric acid
InChI Formula InChI=1S/C5H10O2/c1-4(2)3-5(6)7/h4H,3H2,1-2H3,(H,6,7)
SMILES CC(C)CC(=O)O
IUPAC Name 3-methylbutanoic acid
Molecular Weight (g/mol) 102.13
Formula Weight 102.0681
Chemical Name or Material Isovaleric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.