CAS RN 504-20-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diisopropylidene Acetone, TRC
CAS: 504-20-1 Molecular Formula: C9H14O Molecular Weight (g/mol): 138.21 Synonym: Diisobutenyl Ketone,Diisopropylidene Acetone,Foron,NSC 38718,NSC 403517,Phorone,sym-Diisopropylidene Acetone,2,6-Dimethyl-2,5-heptadien-4-one SMILES: O=C(/C=C(C)\C)/C=C(C)\C