accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 504-63-2

IUPAC Name: propane-1,3-diol
Synonyms: 1,3-propandiol β-propylene glycol 1,3-propylenediol 1,3-propylene glycol ω-propanediol
Molecular Weight (g/mol): 76.1
Molecular Formula: C3H8O2
InChi Key: YPFDHNVEDLHUCE-UHFFFAOYSA-N
SMILES: OCCCO
Molecular Weight (g/mol) 
  • (4)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (4)
Percent Purity 
  • (4)
  • (3)
Physical Form 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (4)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (4)
  • (3)
  • (1)
  • (4)