accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNNNH2

CAS RN 50593-30-1

IUPAC Name: 2-methyl-2H-indazol-6-amine
Synonyms: 2-methylindazol-6-amine
Molecular Weight (g/mol): 147.18
Molecular Formula: C8H9N3
InChi Key: MHCWLERQNFATHZ-UHFFFAOYSA-N
SMILES: CN1C=C2C=CC(N)=CC2=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results