accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNNNNNNNNN

CAS RN 510758-28-8

IUPAC Name: tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
Synonyms: tris(benzyltriazolylmethyl)amine
Molecular Weight (g/mol): 530.64
Molecular Formula: C30H30N10
InChi Key: WKGZJBVXZWCZQC-UHFFFAOYSA-N
SMILES: C(N(CC1=CN(CC2=CC=CC=C2)N=N1)CC1=CN(CC2=CC=CC=C2)N=N1)C1=CN(CC2=CC=CC=C2)N=N1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)