accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5108-96-3

IUPAC Name: pyrrolidine-1-carbodithioic acid amine
Synonyms: pyrrolidine dithiocarbamate amine
Molecular Weight (g/mol): 164.29
Molecular Formula: C5H12N2S2
InChi Key: MDDIUTVUBYEEEM-UHFFFAOYSA-N
SMILES: N.SC(=S)N1CCCC1
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results