missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrrolidinecarbodithioic acid ammonium salt, 98%

Product Code. 11470158
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470158 100 g 100g
11460158 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470158 Supplier Thermo Scientific Alfa Aesar Supplier No. B23731.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5108-96-3 | C5H8NS2- | 146.246 g/mol

1-Pyrrolidinecarbodithioic acid ammonium salt is useful for biochemical applications including metal chelation. It is used to prevent the induction of nitric oxide synthase. It is also essential to inhibit apoptosis in leukemia HL-60 cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrrolidinecarbodithioic acid ammonium salt is useful for biochemical applications including metal chelation. It is used to prevent the induction of nitric oxide synthase. It is also essential to inhibit apoptosis in leukemia HL-60 cells.

Solubility
Soluble in water.

Notes
Hygroscopic. Air and light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5108-96-3
Molecular Formula C5H8NS2-
Molecular Weight (g/mol) 146.246
MDL Number MFCD00012720
InChI Key VSWDORGPIHIGNW-UHFFFAOYSA-M
Synonym ammonium pyrrolidyldithiocarbamate, apdc, ammonium 1-pyrrolidinecarbodithioate, ammonium tetramethylenedithiocarbamate, ammonium pyrrolidine-1-carbodithioate, ammonium 1-pyrrolidinyldithiocarbamate, ammonium 1-pyrrolidinedithiocarboxylate, 1-pyrrolidinecarbodithioic acid, ammonium salt, 1-pyrrolidinecarbodithioic acid ammonium salt, pyrrolidinedithiocarbamate ammonium
PubChem CID 4311638
IUPAC Name pyrrolidine-1-carbodithioate
SMILES C1CCN(C1)C(=S)[S-]

Specifications

Melting Point 150°C to 155°C
Quantity 100 g
Beilstein 3730472
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 164.29
Percent Purity 98%
Chemical Name or Material 1-Pyrrolidinecarbodithioic acid ammonium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.