accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NCOCH3

CAS RN 51163-27-0

IUPAC Name: 4-isocyanato-1,2-dimethylbenzene
Synonyms: 3,4-dimethylphenyl isocyanate 3,4-dimethyl phenylisocyanate
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: AYCDBMRVKSXYKW-UHFFFAOYSA-N
SMILES: CC1=C(C)C=C(C=C1)N=C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)