missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylphenyl isocyanate, 97%

Product Code. 11392837
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392837 1 g 1g
11302847 5 g 5g
11312847 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11392837 Supplier Thermo Scientific Alfa Aesar Supplier No. L15998.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 51163-27-0 | C9H9NO | 147.18 g/mol

3,4-Dimethylphenyl isocyanate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylphenyl isocyanate is used as a pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51163-27-0
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00013867
InChI Key AYCDBMRVKSXYKW-UHFFFAOYSA-N
Synonym 3,4-dimethylphenyl isocyanate, 3,4-dimethylphenylisocyanate, benzene, 4-isocyanato-1,2-dimethyl, 3,4-dimethyl phenylisocyanate, 3,4-dimethylbenzenisocyanate, acmc-1aump, ksc265g2l, 3,4-dimethyl phenyl isocyanate, 1-isocyanato-3,4-dimethyl-benzene, 4-isocyanato-1,2-dimethyl benzene
PubChem CID 4389663
ChEBI CHEBI:60098
IUPAC Name 4-isocyanato-1,2-dimethylbenzene
SMILES CC1=C(C)C=C(C=C1)N=C=O

Specifications

Density 1.051
Quantity 1 g
Refractive Index 1.538
UN Number UN2206
Beilstein 775149
Boiling Point 85°C (2 mmHg)
Solubility Information Hydrolyzes in water.
Formula Weight 147.18
Percent Purity 97%
Chemical Name or Material 3,4-Dimethylphenyl isocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.