accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CO(S)OHO

CAS RN 51268-87-2

IUPAC Name: 3-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]propan-1-ol
Synonyms: 3-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]propan-1-ol
Molecular Weight (g/mol): 160.21
Molecular Formula: C8H16O3
InChi Key: BSLDYXOPGSOQGZ-ZETCQYMHSA-N
SMILES: CC1(C)OC[C@H](CCCO)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results