accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 513-86-0

IUPAC Name: 3-hydroxybutan-2-one
Synonyms: acetoin 2-acetoin 3H-2B butanone acetoin (natural) (R)-2-acetoin 2,3-butanolone (R)-dimethylketol dimethylketol 3h-2b butanone acethoin
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: ROWKJAVDOGWPAT-UHFFFAOYNA-N
SMILES: CC(O)C(C)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results