accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5132-30-9

OONNOO

CAS RN 5132-30-9

IUPAC Name: 1-[2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl]-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-[2-(2,5-dioxopyrrol-1-yl)ethyl]pyrrole-2,5-dione
Molecular Weight (g/mol): 220.18
Molecular Formula: C10H8N2O4
InChi Key: PUKLCKVOVCZYKF-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1CCN1C(=O)C=CC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results