missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bismaleimidoethane, Thermo Scientific Chemicals

Product Code. 17281192 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17281192 500 mg 500mg
17271192 100 mg 100mg
17261192 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17281192 Supplier Thermo Scientific Acros Supplier No. 466065000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5132-30-9
Molecular Formula C10H8N2O4
Molecular Weight (g/mol) 220.18
InChI Key PUKLCKVOVCZYKF-UHFFFAOYSA-N
IUPAC Name 1-[2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl]-2,5-dihydro-1H-pyrrole-2,5-dione
SMILES O=C1C=CC(=O)N1CCN1C(=O)C=CC1=O

Specifications

Melting Point 195.0°C
Quantity 500 mg
Formula Weight 220.18
Chemical Name or Material 1,2-Bismaleimidoethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.